6-Chloro-4-quinazolone 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloroquinazolin-4-ol

A221975-100G A221975-10G A221975-1G A221975-250MG A221975-5G
77097-972EA 206.88 USD
77097-972 77097-974 77097-976 77097-978 77097-980
6-Chloro-4-quinazolone 97%
6-Chloro-4-quinazolone

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O
MW: 180.5900 g/mol
MDL Number: MFCD01686365
CAS Number: 16064-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR