Ethyl-2-oxopyrrolidine-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-oxo-pyrrolidine-3-carboxylate

A220326-100MG A220326-1G A220326-250MG A220326-5G
77097-950EA 31.83 USD
77097-950 77097-952 77097-954 77097-956
Ethyl-2-oxopyrrolidine-3-carboxylate 95%
Ethyl-2-oxopyrrolidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃
MW: 157.17 g/mol
MDL Number: MFCD06796534
CAS Number: 36821-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR