1-(5-Bromo-4-methyl-1H-pyrrol-3-yl)ethanone 97%

Supplier: Ambeed
Warning

A219755-100MG A219755-1G A219755-250MG
77097-944EA 30.77 USD
77097-944 77097-946 77097-948
1-(5-Bromo-4-methyl-1H-pyrrol-3-yl)ethanone 97%
1-(5-Bromo-4-methyl-1H-pyrrol-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.05 g/mol
MDL Number: MFCD11109865
CAS Number: 202286-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR