1-(2-Methoxyethyl)-4-nitro-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
A219038-1G
A219038-250MG
77097-914EA
51
USD
77097-914
77097-916
1-(2-Methoxyethyl)-4-nitro-1H-pyrazole 97%
1-(2-Methoxyethyl)-4-nitro-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₆H₉N₃O₃ |
MDL Number:
MFCD02755678 CAS Number: 948570-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...