2-(5-Amino-1,2,4-thiadiazol-3-yl)-2-(methoxyimino)acetic acid 98%

Supplier: Ambeed
Danger

A218941-1G A218941-250MG A218941-5G
77097-908EA 7.43 USD
77097-908 77097-910 77097-912
2-(5-Amino-1,2,4-thiadiazol-3-yl)-2-(methoxyimino)acetic acid 98%
2-(5-Amino-1,2,4-thiadiazol-3-yl)-2-(methoxyimino)acetic acid

Precaución: for Research Use Only

Formula: C₅H₆N₄O₃S
MW: 202.19 g/mol
MDL Number: MFCD08444061
CAS Number: 72217-12-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR