3-Fluoro-4-(methylsulfonyl)phenylboronic Acid Pinacol Ester 95+%

Supplier: Ambeed
Warning

A218782-100MG A218782-1G A218782-250MG
77097-902EA 18 USD
77097-902 77097-904 77097-906
3-Fluoro-4-(methylsulfonyl)phenylboronic Acid Pinacol Ester 95+%
3-Fluoro-4-(methylsulfonyl)phenylboronic Acid Pinacol Ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₄S
MDL Number: MFCD18760182
CAS Number: 648904-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR