4,5-Dihydroxy-m-anisic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-O-Methylgallic acid, 3,4-Dihydroxy-5-methoxybenzoic acid, 3,4-Dihydroxy-5-Methoxy-Benzoic Acid, 3,4-Dihydroxy-5-methoxybenzenecarboxylic acid

A218322-10G A218322-1G A218322-250MG A218322-5G A218322-25G
77097-894EA 177 USD
77097-894 77097-896 77097-898 77097-900 77685-591
4,5-Dihydroxy-m-anisic acid 97%
4,5-Dihydroxy-m-anisic acid

Precaución: for Research Use Only

Formula: C₈H₈O₅
MW: 184.15 g/mol
MDL Number: MFCD00016518
CAS Number: 3934-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR