Fmoc-D-Glu-OH 95%

Supplier: Ambeed
Warning

Synonyms: Nα-Fmoc-L-glutamic acid, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic acid, N-Fmoc-D-glutamic Acid

A217833-100G A217833-10G A217833-1G A217833-25G
77097-874EA 120 USD
77097-874 77097-876 77097-878 77097-880 77097-882
Fmoc-D-Glu-OH 95%
Fmoc-D-Glu-OH

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₆
MDL Number: MFCD09037362
CAS Number: 104091-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR