3-Chloro-5-(trifluoromethoxy)bromobenzene 95%

Supplier: Ambeed
Warning

A217755-5G A217755-1G
77097-872EA 44 USD
77097-872 77901-515
3-Chloro-5-(trifluoromethoxy)bromobenzene 95%
3-Chloro-5-(trifluoromethoxy)bromobenzene

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O
MDL Number: MFCD11856023
CAS Number: 1417567-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR