(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-fluoro-1H-indol-3-yl)propanoic acid 98%
Supplier: Ambeed
|
Warning
|
A217321-100MG
A217321-1G
A217321-250MG
A217321-5G
77097-856EA
20
USD
77097-856
77097-858
77097-860
77097-862
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-fluoro-1H-indol-3-yl)propanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-fluoro-1H-indol-3-yl)propanoic acid
Precaución: for Research Use Only
Formula:
C₂₆H₂₁FN₂O₄ |
MDL Number:
MFCD24842544 CAS Number: 908846-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...