2,3-Dichloro-5-nitropyridine 98%

Supplier: Ambeed
Danger

A217190-10G A217190-1G A217190-25G A217190-5G
77097-848EA 16 USD
77097-848 77097-850 77097-852 77097-854
2,3-Dichloro-5-nitropyridine 98%
2,3-Dichloro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₂O₂
MDL Number: MFCD03840432
CAS Number: 22353-40-8
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR