5-Amino-3-bromo-2-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-6-methoxypyridin-3-amine

A216251-100G A216251-10G A216251-1G A216251-250MG A216251-25G A216251-5G
77097-830EA 250 USD
77097-830 77097-832 77097-834 77097-836 77097-838 77097-840
5-Amino-3-bromo-2-methoxypyridine 98%
5-Amino-3-bromo-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MDL Number: MFCD06659518
CAS Number: 53242-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR