3-(4-((2,4-Bis(trifluoromethyl)benzyl)oxy)-3-methoxyphenyl)-2-cyano-N-(5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl)acrylamide 98%
Supplier: Ambeed
|
Warning
|
A216057-10MG
A216057-1MG
A216057-50MG
A216057-5MG
77097-822EA
88.99
USD
77097-822
77097-824
77097-826
77097-828
3-(4-((2,4-Bis(trifluoromethyl)benzyl)oxy)-3-methoxyphenyl)-2-cyano-N-(5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl)acrylamide 98%
3-(4-((2,4-Bis(trifluoromethyl)benzyl)oxy)-3-methoxyphenyl)-2-cyano-N-(5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl)acrylamide
Precaución: for Research Use Only
Formula:
C₂₃H₁₃F₉N₄O₃S |
MDL Number:
MFCD08277031 CAS Number: 725247-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...