2-Bromo-4-nitrophenol 98%

Supplier: Ambeed
Danger

Synonyms: 1-Bromo-2-hydroxy-5-nitrobenzene

A215987-10G A215987-1G A215987-25G
77097-818EA 12 USD
77097-818 77097-820 77685-582
2-Bromo-4-nitrophenol 98%
2-Bromo-4-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄BrNO₃
MDL Number: MFCD06656567
CAS Number: 5847-59-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR