2-((1H-Pyrrolo[2,3-b]pyridin-5-yl)oxy)-4-(4-((4'-chloro-5,5-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)benzoic acid 95%

Supplier: Ambeed
Warning

A215704-100MG A215704-1G A215704-250MG A215704-25G A215704-5G
77097-810EA 28.65 USD
77097-810 77097-812 77097-814 77097-816 77685-580
2-((1H-Pyrrolo[2,3-b]pyridin-5-yl)oxy)-4-(4-((4'-chloro-5,5-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)benzoic acid 95%
2-((1H-Pyrrolo[2,3-b]pyridin-5-yl)oxy)-4-(4-((4'-chloro-5,5-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₃₃H₃₅ClN₄O₃
MW: 571.10 g/mol
MDL Number: MFCD28129979
CAS Number: 1235865-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR