4-Bromo-3-chloroaniline 98%

Supplier: Ambeed
Warning

A214515-100G A214515-10G A214515-25G A214515-500G A214515-5G
77097-796EA 66 USD
77097-796 77097-798 77097-800 77097-802 77568-068
4-Bromo-3-chloroaniline 98%
4-Bromo-3-chloroaniline

Precaución: For research use only.

Formula: C₆H₅BrClN
MDL Number: MFCD00041311
CAS Number: 21402-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR