4-Bromo-8-methylquinoline 98%

Supplier: Ambeed
Warning

A213520-100MG A213520-1G A213520-250MG A213520-5G
77097-774EA 44.56 USD
77097-774 77097-776 77097-778 77097-780
4-Bromo-8-methylquinoline 98%
4-Bromo-8-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.0800 g/mol
MDL Number: MFCD08063210
CAS Number: 36075-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR