N-Carbobenzoxy-6-aminohexanoic acid 98%

Supplier: Ambeed
Warning

A213499-25G A213499-5G A213499-100G A213499-10G A213499-500G
77097-770EA 32 USD
77097-770 77097-772 77612-096 77612-098 77685-570
N-Carbobenzoxy-6-aminohexanoic acid 98%
N-Carbobenzoxy-6-aminohexanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MDL Number: MFCD00004423
CAS Number: 1947-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR