3-Bromo-α,α,α-trifluorotoluene 98%

Supplier: Ambeed
Danger

A213195-10G A213195-25G A213195-5G A213195-100G
77097-758EA 6 USD
77097-758 77097-760 77097-762 77685-567
3-Bromo-α,α,α-trifluorotoluene 98%
3-Bromo-α,α,α-trifluorotoluene

Precaución: for Research Use Only

Formula: C₇H₄BrF₃
MDL Number: MFCD00000380
CAS Number: 401-78-5
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR