Methyl 2-(3-bromo-4-hydroxyphenyl)acetate 95%

Supplier: Ambeed
Warning

A213006-100MG A213006-1G A213006-250MG A213006-5G
77097-746EA 24.41 USD
77097-746 77097-748 77097-750 77685-565
Methyl 2-(3-bromo-4-hydroxyphenyl)acetate 95%
Methyl 2-(3-bromo-4-hydroxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
MDL Number: MFCD16293381
CAS Number: 34918-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR