N,N'-Bis(2-hydroxyethyl)ethylenediamine 98%
Supplier: Ambeed
|
Danger
|
A212235-25G
A212235-5G
77097-714EA
29
USD
77097-714
77097-716
N,N'-Bis(2-hydroxyethyl)ethylenediamine 98%
N,N'-Bis(2-hydroxyethyl)ethylenediamine
Precaución: for Research Use Only
Formula:
C₆H₁₆N₂O₂ |
MDL Number:
MFCD00002842 CAS Number: 4439-20-7 UN: 3259 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...