Fmoc-3-iodo-D-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Phe(3-I)-OH

A212143-10G A212143-1G A212143-5G
77097-704EA 383 USD
77097-704 77097-706 77097-708
Fmoc-3-iodo-D-phenylalanine 97%
Fmoc-3-iodo-D-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₀INO₄
MW: 513.34 g/mol
MDL Number: MFCD03840402
CAS Number: 478183-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR