2-Cyano-5-(trifluoromethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

A211200-100MG A211200-1G A211200-250MG A211200-5G
77097-680EA 37.14 USD
77097-680 77097-682 77097-684 77097-686
2-Cyano-5-(trifluoromethyl)phenylboronic acid 98%
2-Cyano-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₅BF₃NO₂
MW: 214.94 g/mol
MDL Number: MFCD10696675
CAS Number: 1375110-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR