7-Bromoquinoline 98%

Supplier: Ambeed
Warning

A210895-100G A210895-10G A210895-1G A210895-25G A210895-5G A210895-500G
77097-670EA 225 USD
77097-670 77097-672 77097-674 77097-676 77097-678 77685-552
7-Bromoquinoline 98%
7-Bromoquinoline

Precaución: for Research Use Only

Formula: C₉H₆BrN
MDL Number: MFCD03695823
CAS Number: 4965-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR