2-Iodo-4-(trifluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

A210423-1G A210423-250MG A210423-5G
77097-654EA 29 USD
77097-654 77097-656 77097-658
2-Iodo-4-(trifluoromethyl)benzoic acid 98%
2-Iodo-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₃IO₂
MDL Number: MFCD06408817
CAS Number: 54507-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR