2-Amino-3-chloro-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A210244-1000G A210244-100G A210244-25G A210244-500G A210244-5G
77097-644EA 239 USD
77097-644 77097-646 77097-648 77097-650 77097-652
2-Amino-3-chloro-5-(trifluoromethyl)pyridine 98%
2-Amino-3-chloro-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄ClF₃N₂
MW: 196.5600 g/mol
MDL Number: MFCD00042154
CAS Number: 79456-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR