4-Hydroxy-6-(trifluoromethyl)pyrimidine 97%

Supplier: Ambeed
Warning

A208860-100G A208860-10G A208860-1G A208860-5G
77097-632EA 323.58 USD
77097-632 77097-634 77097-636 77685-539
4-Hydroxy-6-(trifluoromethyl)pyrimidine 97%
4-Hydroxy-6-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₃F₃N₂O
MW: 164.09 g/mol
MDL Number: MFCD00192530
CAS Number: 1546-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR