5-Fluoro-2-methylbenzyl bromide 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Bromomethyl)-4-fluoro-1-methylbenzene

A208699-10G A208699-1G A208699-25G A208699-5G
77097-624EA 82 USD
77097-624 77097-626 77097-628 77097-630
5-Fluoro-2-methylbenzyl bromide 98%
5-Fluoro-2-methylbenzyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrF
MW: 203.05
MDL Number: MFCD01631528
CAS Number: 261951-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR