2-(2-Bromo-5-(trifluoromethyl)phenyl)acetic acid 95%

Supplier: Ambeed
Warning

A208004-100MG A208004-1G
77097-614EA 51.99 USD
77097-614 77097-616
2-(2-Bromo-5-(trifluoromethyl)phenyl)acetic acid 95%
2-(2-Bromo-5-(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₂
MW: 283.04 g/mol
MDL Number: MFCD13194216
CAS Number: 1214327-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR