3'-Bromo-2'-fluoroacetophenone 98%

Supplier: Ambeed
Warning

A207550-100G A207550-10G A207550-1G A207550-250MG A207550-25G A207550-5G
77097-594EA 692 USD
77097-594 77097-596 77097-598 77097-600 77097-602 77097-604
3'-Bromo-2'-fluoroacetophenone 98%
3'-Bromo-2'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD09264507
CAS Number: 161957-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR