3-Chlorobenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: (3-Chlorophenyl)methanamine

A207391-10G A207391-1G A207391-25G A207391-5G
77097-586EA 23 USD
77097-586 77097-588 77097-590 77097-592
3-Chlorobenzylamine 98%
3-Chlorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₈ClN
MW: 141.6
Boiling Pt: 40
Density: 1.159
MDL Number: MFCD00040752
CAS Number: 4152-90-3
UN: 2735
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR