4-Chloro-2-methylphenylboronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Chloro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Chloro-2-methylphenylboronic acid pinacol ester

A207025-1G A207025-250MG A207025-5G
77097-580EA 15.92 USD
77097-580 77097-582 77097-584
4-Chloro-2-methylphenylboronic acid, pinacol ester 97%
4-Chloro-2-methylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BClO₂
MW: 252.545 g/mol
MDL Number: MFCD18729895
CAS Number: 1030832-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR