4,5-Dibromo-1-methyl-1,2,3-triazole 95%

Supplier: Ambeed
Warning

A207001-250MG A207001-5G A207001-25G
77097-572EA 8 USD
77097-572 77097-574 77685-537
4,5-Dibromo-1-methyl-1,2,3-triazole 95%
4,5-Dibromo-1-methyl-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₃H₃Br₂N₃
MDL Number: MFCD00499501
CAS Number: 25537-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR