(4S)-4-Benzyl-1,3-thiazolidine-2-thione 97%

Supplier: Ambeed
Warning

Synonyms: (S)-4-Benzylthiazolidine-2-thione, (4S)-4-(Phenylmethyl)-2-thioxo-1,3-thiazolidine, (4S)-4-Benzyl-2-thioxo-1,3-thiazolidine

A206484-1G A206484-250MG A206484-25G A206484-5G
77097-556EA 49.87 USD
77097-556 77097-558 77097-560 77097-562
(4S)-4-Benzyl-1,3-thiazolidine-2-thione 97%
(4S)-4-Benzyl-1,3-thiazolidine-2-thione

Precaución: for Research Use Only

Formula: C₁₀H₁₁NS₂
MW: 209.331 g/mol
MDL Number: MFCD06658216
CAS Number: 171877-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR