3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine 95%

Supplier: Ambeed
Warning

A206138-100MG A206138-1G A206138-250MG
77097-544EA 150.65 USD
77097-544 77097-546 77097-548
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine 95%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₂H₁₉BN₂O₂
MW: 234.10 g/mol
MDL Number: MFCD22494032
CAS Number: 873663-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR