Ifosfamide 98%

Supplier: Ambeed
Danger

Synonyms: Ifex, 3-(2-Chloroethyl)-2-((2-chloroethyl)amino)-1,3,2-oxazaphosphinane 2-oxide, N,3-Bis(2-chloroethyl)tetrahydro-2H-1,3,2-oxazaphosphorin-2-amine-2-oxide

A205362-1G A205362-50MG A205362-25G A205362-5G
77097-524EA 27.59 USD
77097-524 77097-526 77685-525 77685-526
Ifosfamide 98%
Ifosfamide

Precaución: for Research Use Only

Formula: C₇H₁₅Cl₂N₂O₂P
MW: 261.09 g/mol
MDL Number: MFCD00057374
CAS Number: 3778-73-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR