4-Bromo-2-nitrophenol 98%

Supplier: Ambeed
Danger

A204748-100G A204748-10G A204748-1G A204748-25G A204748-500G A204748-5G
77097-508EA 84 USD
77097-508 77097-510 77097-512 77097-514 77097-516 77097-518
4-Bromo-2-nitrophenol 98%
4-Bromo-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄BrNO₃
MDL Number: MFCD00082540
CAS Number: 7693-52-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR