2-Amino-4-bromo-3-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-6-carboxy-2-fluoroaniline, 2-amino-4-bromo-3-fluorobenzoic acid, 4-Bromo-3-fluoroanthranilic acid

A203784-100G A203784-100MG A203784-10G A203784-1G A203784-250MG A203784-25G A203784-5G
77097-484EA 415 USD
77097-484 77097-486 77097-488 77097-490 77097-492 77097-494 77097-496
2-Amino-4-bromo-3-fluorobenzoic acid 97%
2-Amino-4-bromo-3-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD27664889
CAS Number: 1416013-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR