Asymmetric dimethylarginine 95%

Supplier: Ambeed
Warning

Synonyms: NG,NG-Dimethylarginine, NG,NG-dimethyl-L-Arginine, Dimethyl-L-arginine, ADMA, (S)-2-Amino-5-(3,3-dimethylguanidino)pentanoic acid

A203396-1G A203396-250MG
77097-474EA 523.03 USD
77097-474 77097-476
Asymmetric dimethylarginine 95%
Asymmetric dimethylarginine

Precaución: for Research Use Only

Formula: C₈H₁₈N₄O₂
MW: 202.25 g/mol
MDL Number: MFCD00038406
CAS Number: 30315-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR