3',4'-Difluoroacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(3,4-Difluorophenyl)ethan-1-one, 1-(3,4-Difluorophenyl)ethanone

A203149-100G A203149-10G A203149-25G A203149-500G A203149-5G
77097-464EA 45.62 USD
77097-464 77097-466 77097-468 77097-470 77097-472
3',4'-Difluoroacetophenone 97%
3',4'-Difluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆F₂O
MW: 156.13 g/mol
MDL Number: MFCD00009891
CAS Number: 369-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR