N-(Heptan-4-yl)benzo[d][1,3]dioxole-5-carboxamide 95%

Supplier: Ambeed
Warning

A203048-100MG A203048-1G A203048-250MG
77097-458EA 48.81 USD
77097-458 77097-460 77097-462
N-(Heptan-4-yl)benzo[d][1,3]dioxole-5-carboxamide 95%
N-(Heptan-4-yl)benzo[d][1,3]dioxole-5-carboxamide

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₃
MW: 263.33 g/mol
MDL Number: MFCD08443801
CAS Number: 745047-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR