2-Chloro-7-cyclopentyl-N,N-dimethyl-7H-pyrrolo[2,3-d]pyrimidine-6-carboxamide 95%
Supplier: Ambeed
|
Warning
|
A202610-1G
A202610-250MG
A202610-5G
77097-432EA
44.56
USD
77097-432
77097-434
77097-436
2-Chloro-7-cyclopentyl-N,N-dimethyl-7H-pyrrolo[2,3-d]pyrimidine-6-carboxamide 95%
2-Chloro-7-cyclopentyl-N,N-dimethyl-7H-pyrrolo[2,3-d]pyrimidine-6-carboxamide
Precaución: for Research Use Only
Formula:
C₁₄H₁₇ClN₄O MW: 292.76 g/mol |
MDL Number:
MFCD20527881 CAS Number: 1211443-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...