1-(2-Chlorophenyl)ethanamine 95%

Supplier: Ambeed
Warning

A200436-1G A200436-250MG
77097-392EA 40 USD
77097-392 77097-394
1-(2-Chlorophenyl)ethanamine 95%
1-(2-Chlorophenyl)ethanamine

Precaución: for Research Use Only

Formula: C₈H₁₀ClN
MDL Number: MFCD05215230
CAS Number: 39959-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR