Ethyl-8-bromo-4-hydroxyquinoline-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 8-bromo-4-hydroxyquinoline-3-carboxylate

A200224-100MG A200224-1G A200224-250MG A200224-5G
77097-384EA 7.43 USD
77097-384 77097-386 77097-388 77097-390
Ethyl-8-bromo-4-hydroxyquinoline-3-carboxylate 97%
Ethyl 8-bromo-4-hydroxyquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO₃
MW: 296.1167 g/mol
MDL Number: MFCD00173380
CAS Number: 35975-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR