3-Fluoro-4-methoxybenzoyl chloride 98%

Supplier: Ambeed
Danger

A198916-1G A198916-25G A198916-5G
77097-368EA 14 USD
77097-368 77097-370 77097-372
3-Fluoro-4-methoxybenzoyl chloride 98%
3-Fluoro-4-methoxybenzoyl chloride

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MDL Number: MFCD03428589
CAS Number: 3907-15-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR