3-Hydrazinylbenzonitrile hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Cyanophenylhydrazine hydrochloride
A198371-100G
A198371-10G
A198371-1G
A198371-25G
A198371-5G
77097-344EA
195
USD
77097-344
77097-346
77097-348
77097-350
77097-352
3-Hydrazinylbenzonitrile hydrochloride 97%
3-Hydrazinylbenzonitrile hydrochloride
Precaución: for Research Use Only
Formula:
C7H8ClN3 MW: 169.6115 g/mol |
MDL Number:
MFCD03094642 CAS Number: 2881-99-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...