Methyl 4-bromo-2-fluoro-6-methylbenzoate 95%

Supplier: Ambeed
Warning

A198310-100MG A198310-5G A198310-1G A198310-25G A198310-250MG
77097-342EA 10 USD
77097-342 77593-280 77611-904 77611-906 77901-542
Methyl 4-bromo-2-fluoro-6-methylbenzoate 95%
Methyl 4-bromo-2-fluoro-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MDL Number: MFCD23703481
CAS Number: 1427409-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR