7-Methoxy-4-(piperazin-1-yl)quinoline 95%

Supplier: Ambeed
Warning

A198266-100MG A198266-1G A198266-250MG
77097-336EA 62.6 USD
77097-336 77097-338 77097-340
7-Methoxy-4-(piperazin-1-yl)quinoline 95%
7-Methoxy-4-(piperazin-1-yl)quinoline

Precaución: for Research Use Only

Formula: C₁₄H₁₇N₃O
MW: 243.304 g/mol
MDL Number: MFCD02179804
CAS Number: 4038-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR