Boc-Dap(Boc)-OH (dicyclohexylammonium) salt 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Dap(Boc)-OH.DCHA, Boc-Dap(Boc)-OH·DCHA
A197993-10G
A197993-1G
A197993-250MG
A197993-25G
A197993-5G
77097-326EA
40
USD
77097-326
77097-328
77097-330
77097-332
77097-334
Boc-Dap(Boc)-OH (dicyclohexylammonium) salt 97%
Boc-Dap(Boc)-OH (dicyclohexylammonium) salt
Precaución: for Research Use Only
Formula:
C₁₃H₂₄N₂O₆·C₁₂H₂₃N MW: 485.666 g/mol |
MDL Number:
MFCD00238311 CAS Number: 201472-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...