Imidazo[1,2-a]pyridine 95%

Supplier: Ambeed
Warning

A197969-10G A197969-1G A197969-25G A197969-5G
77097-316EA 55.17 USD
77097-316 77097-318 77097-320 77097-322
Imidazo[1,2-a]pyridine 95%
Imidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₆N₂
MW: 118.1359 g/mol
MDL Number: MFCD00005553
CAS Number: 274-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR